Learn More
Medchemexpress LLC Cyclopentaneacetic acid, 3-oxo-2-(2Z)-2-penten-1-yl-, (1R,2R)-rel- | 77026-92-7 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY122464100MG
Encompass_Preferred
(±)-Jasmonic acid is the racemic form of jasmonic acid, a plant hormone and growth regulator that modulates defense responses to wounding, herbivory, and pathogen attack. It is used in plant physiology and signaling research to study stress responses, chlorophyll modulation, and seedling growth.
  • Racemic (±) form of jasmonic acid.
  • Used to activate plant defense and stress response pathways.
  • High purity (99.85%) for reliable experimental results.
  • Available in small research quantities, including 100 mg.
  • Colorless to light yellow solution; store at -20 °C for up to 2 years.
Catalog No. 50-002-32645