Learn More
Medchemexpress LLC Cyclopentanepropanoic acid, α-[(9H-fluoren-9-ylmethoxy)carbonyl]amino | 371770-32-0 | 99.9% | 379.45 g/mol | C23H25NO4 | 1 G

Supplier: Medchemexpress LLC HYW0419831G
Fmoc-Cpa-OH is an N-Fmoc protected cyclopentanepropanoic acid used as a building block in peptide synthesis and other peptide research applications. It is supplied as the free acid with an Fmoc protecting group on the amine, and batch-specific analytical documentation (certificate of analysis) is provided.
- Contains both amino and carboxyl functional groups
- N-Fmoc protected amine for peptide coupling
- Used as a noncanonical amino acid building block in peptide synthesis
- High purity with batch certificate of analysis available
- Soluble in common organic solvents for coupling reactions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.