Learn More
Medchemexpress LLC Cyclopenthiazide | 742-20-1 | 98.4% | 379.88 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10954210MM/1ML
Encompass_Preferred
Cyclopenthiazide is a benzothiadiazine diuretic with antihypertensive properties. It functions by inhibiting the reabsorption of sodium chloride and water in the distal renal tubules, thereby increasing the excretory capacity of the rat kidney.
  • Functions as a benzothiadiazine diuretic.
  • Possesses antihypertensive properties.
  • Inhibits the reabsorption of sodium chloride and water at the distal renal tubules.
  • Increases the excretory capacity of the rat kidney.
Catalog No. 50-003-46030