Learn More
Medchemexpress LLC Cyclophilin B Antibody (YA787) (PBS only) | 24 kDa | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8063850UL
Encompass_Preferred
Cyclophilin B Antibody (YA787) is a mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Cyclophilin B. This protein is a cyclosporine-binding protein primarily located in the endoplasmic reticulum, involved in the secretory pathway and released into biological fluids. It can bind to T- and B-lymphocytes and may regulate cyclosporine A-mediated immunosuppression.
  • Mouse-derived and non-conjugated IgG1 monoclonal antibody
  • Targets cyclophilin B
  • Reactivity across human, rat, and mouse species
  • Supplied in PBS, pH 7.4 formulation
  • Affinity purified for high quality
  • Stable for 1 year when stored at -20°C
  • Suitable for Western Blot (WB) and Immunohistochemistry (IHC-P) applications
Catalog No. 50-003-95509