Learn More
Medchemexpress LLC Cyclophilin F/PPIF P 5ug
SDP
Supplier:  Medchemexpress LLC HYP74211A5UG
Encompass_Preferred
Cyclophilin F/PPIF protein is a peptidyl prolyl cis-trans isomerase (PPIase) that catalyzes the isomerization of proline imide peptide bonds and may contribute to protein folding It critically regulates the mitochondrial permeability transition pore (mPTP) affecting cell apoptosis or necrosis Cyclophilin F/PPIF Protein Human (HEK293 His) is the recombinant human-derived Cyclophilin F/PPIF protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-78790