Learn More
Medchemexpress LLC Cyclophosphamide | 50-18-0 | 99.9% | 261.09 | 1 G
SDP
Supplier:  Medchemexpress LLC HY174201G
Encompass_Preferred
Cyclophosphamide is a synthetic alkylating agent chemically related to nitrogen mustards with antineoplastic activity and is an immunosuppressant. It is for research use only. This agent is a commonly used cross-linking agent that requires metabolic activation, with the active metabolite being 4-Hydroperoxycyclophosphamide.
  • Synthetic alkylating agent
  • Immunosuppressant
  • Requires metabolic activation
  • Target: DNA alkylator
  • Can be used to create models of premature ovarian failure, bone marrow suppression, and immunosuppression
Catalog No. 50-003-84023