Learn More
Selleck Chemical LLC Cyclophosphamide S1217-100mg
SDP
Supplier:  Selleck Chemical LLC S1217100MG
Encompass_Preferred
Cyclophosphamide(NSC 26271 Endoxan Cytoxan Neosar Procytox Revimmune Cytophosphane) is a nitrogen mustard alkylating agent from the oxazophorines group
Catalog No. 50-310-8268