Learn More
TARGETMOL CHEMICALS INC CYCLOPIAZONIC ACID 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T150275MG
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cyclopiazonic acid (CPA) is a neurotoxic secondary metabolite (SM) made by A. flavus. Cyclopiazonic acid (CPA) is an endoplasmic reticulum calcium ATPase (Ca2+ATPase; SERCA)nanomolar inhibitor and a potent inducer of cell death in plants. purity: 99%
Catalog No. 50-258-1257