Learn More
Medchemexpress LLC Cyclopropanecarboxylic acid, 2-(3-pyridinyl)-, (1R,2R)- | 1423769-08-7 | 99.5% | 163.18 g/mol | C9H9NO2 | 100 MG

Supplier: Medchemexpress LLC HYW063971100MG
(1R,2R)-2-(Pyridin-3-yl)cyclopropane-1-carboxylic acid is a chiral cyclopropane-containing carboxylic acid used as an intermediate in medicinal chemistry and organic synthesis. It is supplied as a light yellow to yellow solid with high chemical and enantiomeric purity and is intended for research use; observe recommended storage to maintain stability.
- appearance: light yellow to yellow solid
- purity (lc-ms): 99.46%
- enantiomeric excess: 99.13%
- molecular weight: 163.18 g/mol
- chemical formula: C9H9NO2
- storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months)
- typical use: intermediate for synthesis of active compounds in medicinal chemistry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.