Learn More
Medchemexpress LLC Cyclopropanecarboxylic acid, 2-[4-[bis(2-methylpropyl)amino]-3-[[[(4-methylphenyl)amino]carbonyl)amino]phenyl]-, (1R,2S)- | 1629125-65-0 | 99.9% | 437.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1876950MG
Encompass_Preferred
BMS-978587 is an indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor, extracted from patent WO2014150677A1, Compound example 1 enantiomer 1. It is for research use only.
  • Inhibits IDO-1 in human IDO1/HEK293 cells
Catalog No. 50-003-85699