Learn More
Medchemexpress LLC Cyclopropanesulfonamide, N-[4-[5-[[methyl[2-(methylamino)ethyl]amino]methyl]-3-pyridinyl]phenyl]- | 2890765-10-1 | C19H26N4O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1522365MG
Encompass_Preferred
PRMT6-IN-3 (compound 25) is a selective PRMT6 inhibitor with an IC50 value of 192 nM. It can induce apoptosis in cancer cells and has anticancer activity.
  • Selective PRMT6 inhibitor with an IC50 value of 192 nM.
  • Induces apoptosis in cancer cells.
  • Possesses anticancer activity.
Catalog No. 50-003-73807