Learn More
Medchemexpress LLC Cyclopropanesulfonyl chloride | 139631-62-2 | 140.59 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002798
Encompass_Preferred
Cyclopropanesulfonyl chloride is an intermediate that can be used to synthesize Compound 1. Compound 1 has anticancer activity against EGFR triple-mutant tumors. It appears as a liquid (Density: 1.38 g/cm³) with a colorless to light yellow color.
  • Intermediate for synthesis applications
  • Can be used to synthesize anticancer compounds
  • Liquid form with colorless to light yellow appearance
  • Targeted for biochemical assay reagents and drug intermediate applications
Catalog No. 50-004-48133