Learn More
Medchemexpress LLC Cyclopropanesulfonyl chloride | 139631-62-2 | MFCD01631933 | 140.59 | 5 G
Supplier: Medchemexpress LLC HYW0027985G
Cyclopropanesulfonyl chloride is an intermediate used in organic synthesis. It can be utilized to synthesize compounds exhibiting anticancer activity against EGFR triple-mutant tumors.
- Used to synthesize compounds with anticancer activity
- Effective against EGFR triple-mutant tumors
- Liquid appearance with colorless to light yellow color
- Soluble in DMSO and corn oil for in vitro and in vivo applications