Learn More
Medchemexpress LLC Cyclopropyl-[4-[3-(1H-imidazol-5-yl)propoxy]phenyl]methanone | 184025-18-1 | MFCD00951610 | >98.0% | 270.33 g/mol | C16H18N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145675MG
Encompass_Preferred
Ciproxifan is a research reagent that functions as a selective histamine H3 receptor antagonist (synonym FUB-359). The listing corresponds to the free base (CAS 184025-18-1) and is provided as a high-purity compound for pharmacology and receptor-binding studies.
  • Selective histamine H3 receptor antagonist for receptor pharmacology studies.
  • Free base form with CAS 184025-18-1.
  • High purity >98.0%.
  • Small research pack: 5 MG.
  • Chemical formula C16H18N2O2; molecular weight 270.33 g/mol.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-21125