Learn More
Abcam Cyclosporin A, PP2B inhibitor, 500MG
Supplier:  Abcam AB120114500MG
Encompass_Preferred
MW 1202.6 Da, Purity >99%. Immunosuppressant. Complexes with cyclophilin to inhibit the phosphatase activity of calcineurin (PP2B). Please see CofA for batch specific MW.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4691