Learn More
Medchemexpress LLC Cyclosporin B | 63775-95-1 | 99.7% | 1188.58 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP19735MG
Encompass_Preferred
Cyclosporin B is a group of nonpolar cyclic oligopeptides with immunosuppressive activity. It is used for the prevention of graft rejection in organ transplantation.
  • Solid appearance
  • White to off-white color
  • Ships at room temperature in continental US
  • Requires sealed storage, away from moisture and light
  • Powder stable for 2 years at -80°C or 1 year at -20°C
  • In solvent stable for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-22886