Learn More
AdipoGen Cyclosporin C (5 mg)
SDP
Supplier:  AdipoGen AGCN20443M005
Encompass_Preferred
Chemical. CAS: 59787-61-0. Formula: C62H111N11O13. MW: 1218.6. Isolated from Fusarium solani. Immunosuppressant. Calcineurin inhibitor with lower activity than Cyclosporin A (AG-CN2-0079). Antifungal. Antitumor.
Catalog No. 50-205-3611