Learn More
Apexbio Technology LLC Cyclosporin D 63775-96-2 25mg
SDP
Supplier:  Apexbio Technology LLC B591525
Encompass_Preferred
Cyclosporin D (CAS 63775-96-2) is a structural analogue of Cyclosporin A with comparatively reduced immunosuppressive activity Mechanistically it weakly inhibits lymphocyte proliferation induced by mitogens such as PHA PWM PMA and Ca 2 and suppresses Ca 2 /calmodulin-dependent phosphorylation of elongation factor 2 (EF-2) in mouse pancreatic cells In preclinical models Cyclosporin D overcomes Adriamycin resistance in multidrug-resistant ovarian cancer cells and inhibits TPA-induced ear edema and alkaline phosphatase activity in mice Additionally it displays inhibitory effects on the growth of malarial parasites both in vitro and in vivo Cyclosporin D serves as a standard in Cyclosporin A quantification and as a research tool in immunology oncology and antiparasitic studies
Catalog No. 50-000-95338