Learn More
Cayman Chemical CyclosporIn H 10mg
SDP
Supplier:  Cayman Chemical 1718210 MG
Encompass_Preferred
A natural cyclic undecapeptide that selectively antagonizes the formyl peptide receptor at concentrations ranging from 0.1 to 10 µM
Catalog No. 50-296-6117