Learn More
Cayman Chemical CyclothIazIde 10mg
SDP
Supplier:  Cayman Chemical 1633510 MG
Encompass_Preferred
A benzothiadiazide that acts as a potentiator of AMPA receptors, positively modulating its response to glutamic acid (EC50 = 3.8 µM); also a negative modulator of GABAA receptors, reversibly inhibiting both evoked and spontaneous inhibitory postsynaptic currents (IC50 = 58 µM)
Catalog No. 50-297-2433