Learn More
Medchemexpress LLC HY-111653 25mg Medchemexpress, CycLuc1 CAS:1247879-16-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11165325MG
Encompass_Preferred
Medchemexpress, HY-111653 25mg CycLuc1 CAS:1247879-16-8 CycLuc1 is a brain penetrant luciferase substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8938