Learn More
Medchemexpress LLC Cycrimine | 77-39-4 | 99.9% | 1 MG

Supplier: Medchemexpress LLC HY1199181MG
Cycrimine is an orally active muscarinic acetylcholine receptor (mAChR) M1 antagonist used in preclinical research to reduce acetylcholine levels in Parkinson disease models. It also exhibits antispasmodic activity and is employed in behavioral and neurological studies.
- Orally active muscarinic acetylcholine receptor (mAChR) M1 antagonist
- Reduces acetylcholine levels in Parkinson disease models
- Shows antispasmodic activity useful in behavioral studies
- High purity: 99.9% suitable for research applications
- Molecular formula C19H29NO; molecular weight 287.44 g/mol
- Available in small research pack sizes such as 1 MG
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.