Learn More
Medchemexpress LLC Cycrimine | 77-39-4 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11991810MG
Encompass_Preferred
Cycrimine is an orally active muscarinic cholinergic receptor (mAChR) M1 antagonist used as a research reagent. It reduces acetylcholine levels in Parkinson's disease models, shows antispasmodic activity, and is used in behavioral and neurological disorder studies.
  • Orally active mAChR M1 antagonist used in in vivo and in vitro research.
  • Reduces acetylcholine levels in Parkinson's disease models.
  • Shows antispasmodic activity relevant to behavioral and neurological studies.
  • Available in small mg-scale quantities suitable for laboratory experiments.
  • Molecular formula C19H29NO and molecular weight 287.44 g·mol⁻¹.
Catalog No. 50-002-34645