Learn More
Medchemexpress LLC λ-Cyhalothrin | 91465-08-6 | 99.9% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0836R
Encompass_Preferred
λ-Cyhalothrin is a highly efficient, broad-spectrum type II synthetic pyrethroid insecticide containing an α-cyano group. It is used to control a wide range of pests by acting as a neurotoxin, targeting sodium channels in the central nervous system. This compound delays sodium channel inactivation, leading to persistent nerve membrane depolarization, calcium ion influx, and neurotransmitter release from rat brain synaptosomes. It also exhibits estrogenic properties.
  • Highly efficient, broad-spectrum type II synthetic pyrethroid.
  • Contains an α-cyano group.
  • Functions as a neurotoxin, targeting sodium channels.
  • Effective in controlling pests such as fleas, cockroaches, flies, and ants.
  • Highly active against malaria-transmitting *Anopheles* species.
  • Exhibits estrogenic properties as a xenoestrogen.
Catalog No. 50-004-06142