Learn More
Medchemexpress LLC Cyjohnphos (2-(Dicyclohexylphosphino)biphenyl) | 247940-06-3 | 350.49 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00284910G
Encompass_Preferred
Cyjohnphos is a biochemical reagent used as a biological material or organic compound for life science-related research. It can be used in combination with a nickel catalyst for the Suzuki-Miyaura arylation of tertiary benzylic acetates with aryl boroxines to produce diaryl and triaryl quaternary stereocenters.
  • Biochemical reagent for life science research
  • Used with a nickel catalyst for Suzuki-Miyaura arylation
  • Produces diaryl and triaryl quaternary stereocenters
Catalog No. 50-004-49136