Learn More
Andwin Scientific CYLINDER BRUSHES
SDP
Supplier:  Andwin Scientific 17041002
Encompass_Preferred
The Mill-Rose cylinder and bottle brush is ideal for washing cylinders, large test tubes and bottles. This product features a radial end that allows this product to efficiently sanitize the desired item. The brush bristles are attached to a twisted wire handle with hanging loop for convenient storage.
Catalog No. NC9434167