Learn More
Chemglass Life Sciences Chemglass CG-8238-50 50 mL Graduated Cylinder, Single Metric Scale (To Contain), Pyrex
SDP
Supplier:  Chemglass Life Sciences CG823850
Encompass_Preferred
Chemglass CG-8238-50 is a 50 mL Pyrex graduated cylinder with a single metric scale calibrated to contain (TC) for measuring and holding liquid volumes in laboratory settings. It is suitable for routine liquid handling and basic volumetric containment tasks.
  • 50 mL capacity.
  • Single metric scale for straightforward volume reading.
  • Calibrated to contain (TC) for accurate holding volumes.
  • Made from Pyrex glass for chemical resistance.
  • Bumper guard included to reduce breakage risk.
Catalog No. NC2959214