Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 100ML
Supplier:  Wilmad Labglass LG5520106
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 100mL; Subdivision: 1mL; Tolerance: 0.50mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-803-715
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.