Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 10ML
Supplier:  Wilmad Labglass LG5520100
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 10mL; Subdivision: 0.1mL; Tolerance: 0.10mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-804-058