Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 500ML
Supplier:  Wilmad Labglass LG5520110
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 500mL; Subdivision: 5mL; Tolerance: 2.00mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-804-833
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.