Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 50ML
Supplier:  Wilmad Labglass LG5520104
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 50mL; Subdivision: 1mL; Tolerance: 0.25mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-803-954