Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 10ML
Supplier:  Wilmad Labglass LG5510102
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 10mL; Subdivisions: 0.2mL; Tolerance: 0.2mL, Approximate Height: 135mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-807-811