Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 50ML
Supplier:  Wilmad Labglass LG5510106
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 50mL; Subdivisions: 1mL; Tolerance: 0.5mL, Approximate Height: 190mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-803-124
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.