Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 5ML
Supplier:  Wilmad Labglass LG5510100
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 5mL; Subdivisions: 0.1mL; Tolerance: 0.1mL, Approximate Height: 110mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-803-303