Learn More
Medchemexpress LLC CYM-5478 | 870762-83-7 | 99.9% | 388.38 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111253
Encompass_Preferred
CYM-5478 is a potent and highly selective S1P2 agonist with an EC50 of 119 nM in a TGFα-shedding assay. It protects neural-derived cell lines against Cisplatin toxicity.
  • Potent and highly selective S1P2 agonist
  • Protects neural-derived cell lines against Cisplatin toxicity
  • Attenuates Cisplatin-induced caspase 3/7 activity
  • Attenuates increase of ROS in C6 cells exposed to Cisplatin
  • Protects against Cisplatin-mediated ototoxicity
  • Protects against loss of hair cell viability in a zebrafish model
Catalog No. 50-003-46815