Learn More
TARGETMOL CHEMICALS INC CYM2503 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T896310MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. CYM2503 is a positive allosteric modulator (PAM) of the GAL2 receptor that potentiates galanin-induced IP1 production in vitro. It has no affinity for the orthosteric galanin binding site of the receptor (as measured by its inability to displace iodinated galanin, or to induce IP1 accumulation on its own). Purity 97.04%
Catalog No. 50-287-7875