Learn More
Medchemexpress LLC HY-108495 5mg Medchemexpress, CYM50308 CAS:1345858-76-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084955MG
Encompass_Preferred
Medchemexpress, HY-108495 5mg CYM50308 CAS:1345858-76-5 CYM50308 is a potent, selective and high affinity sphingosine-1-phosphate receptor 4 (S1P4-R) agonist with an EC50 of 56 nM. CYM50308 displays 37-fold more selective for S1P4-R than S1P5-R. CYM50308 has no activity at S1P1-R, S1P2-R and S1P3-R subtypes at concentrations up to 25 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8561