Learn More
TARGETMOL CHEMICALS INC CYM5181 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T311495MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CYM5181 is related to the body's immune system and can be used to study multiple sclerosis transplant rejection and adult respiratory distress syndrome. purity: 98%
Catalog No. 50-258-0378