Learn More
Medchemexpress LLC Cym5442 | 1094042-01-9 | 98.8% | 409.48 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1096810MG
Encompass_Preferred
CYM5442 is a potent, highly-selective, and orally active sphingosine 1-phosphate (S1P1) receptor agonist with an EC50 of 1.35 nM. It is inactive against S1P2, S1P3, S1P4, and S1P5. This compound activates S1P1-dependent p42/p44-MAPK phosphorylation, exerts retinal neuroprotection, and can easily penetrate the central nervous system (CNS).
  • Potent and highly-selective S1P1 receptor agonist
  • Orally active
  • Inactive against S1P2, S1P3, S1P4, and S1P5
  • Activates S1P1-dependent p42/p44-MAPK phosphorylation
  • Exerts retinal neuroprotection
  • Easily penetrates the central nervous system (CNS)
Catalog No. 50-003-45433