Learn More
Selleck Chemical LLC CYN154806 P1236-5MG
SDP
Supplier:  Selleck Chemical LLC P12365MG
Encompass_Preferred
CYN154806 a cyclic octapeptide is a potent and selective somatostatin SST2 receptor antagonist with pIC50 values of 8 58 5 41 6 07 5 76 and 6 48 for human recombinant SST2 SST1 SST3 SST4 and SST5 receptors respectively
Catalog No. 50-000-40070