Learn More
Selleck Chemical LLC Cynarin S3301-1mg
SDP
Supplier:  Selleck Chemical LLC S33011MG
Encompass_Preferred
Cynarin (Cynarine) is an artichoke phytochemical that possesses a variety of pharmacological features including free-radical scavenging antioxidant antihistamic and antiviral activities Cynarin blocks the interaction between the CD28 of T-cell receptor and CD80 of antigen presenting cells Cynarin triggers Nrf2 nuclear translocation restores the balance of glutathione (GSH) and reactive oxygen species (ROS) and inhibits mitochondrial depolarization
Catalog No. 50-311-0463