Learn More
Cayman Chemical Cyno-myracrylamIde 1mg
SDP
Supplier:  Cayman Chemical 351551 MG
Encompass_Preferred
A zDHHC20 inhibitor (IC50 = 1.35 µM in a cell-free assay); selectively inhibits zDHHC20 over APT-1 and APT-2 at 50 µM; inhibits S-acylation of Legionella E3 ligase GobX, MyD88, and Ras, which are substrates of zDHHC20, zDHHC9, and zDHHC6, respectively, in HEK293T cells expressing recombinant Legionella GobX, recombinant human MyD88, or endogenous Ras; is not toxic to HEK293T cells at 40 µM
Catalog No. 50-291-0381