Learn More
ACROBiosystems Cynomolgus DLL3 Protein, His Tag
Supplier:  ACROBiosystems DL3C52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus DLL3, His Tag (DL3-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # A0A2K5WSR4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4901