Learn More
Sino Biological Recombinant Cynomolgus DPEP1 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Supplier:  Sino Biological 91070C49HB20
Encompass_Preferred
A DNA sequence encoding the Cynomolgus DPEP1 (G7PZZ9) (Met1-Ser385) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3145