Learn More
ACROBiosystems Cynomolgus Fc gamma RIIB / CD32b Protein, His Tag
Supplier:  ACROBiosystems CDBC5224100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus CD32b, His Tag (CDB-C5224) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 224 (Accession # Q8SPW3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22 kDa.
Catalog No. 50-205-4995