Learn More
ACROBiosystems Cynomolgus IL-11 Protein, His Tag
Supplier:  ACROBiosystems IL1C5243100UG
Encompass_Preferred
Cynomolgus IL-11 Protein, His Tag (IL1-C5243) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Accession # P20808).
Catalog No. 50-233-0968