Learn More
Sino Biological Cynomolgus KLRF1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90165C01H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus KLRF1(Q8MI05) (Val66-Tyr231) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-2433