Learn More
Kingfisher Biotech Inc Cynomolgus Monkey M-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1612Y025
Encompass
The Cynomolgus Monkey M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey M-CSF Specifications: (Molecular Weight: 18.4 kDa) (Amino Acid Sequence: EEVSEYCSHM IGSGHLQSLQ RLIDSQMETS CQITFEFVDQ EQLKDPVCYL KKAFLLVQDI MEDTMRFRDN TPNAIAIVQL QELSLRLKSC FTKDYEEHDK ACVRTFYETP LQLLEKVKNV FNETKNLLDK DWNIFSKNCN NSFAECSGQG HERQREGS (158)) (Gene ID: 102141631). For research use only. Made in the USA
Catalog No. 50-253-3070