Learn More
Cayman Chemical Cynomolgus Monkey Pregn X 3x32
SDP
Supplier:  Cayman Chemical 214033X32 WELL
Encompass_Preferred
cPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. C07001-32, C07001]
Catalog No. 50-296-4376