Learn More
Sino Biological Cynomolgus / Rhesus CD64/FCGR1  Protein (His Tag)
Supplier:  Sino Biological 91073C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus, Rhesus FCGR1 (F7BDL2) (Gln18-Pro288) was expressed with a polyhistidine tag at the C-terminus. The sequences of A0A2K5U901 (Cynomolgus) (Gln33–Pro305) and F7BDL2 (Rhesus macaque) (Gln18–Pro288) are identical.
Catalog No. 50-299-3146